Product Details

SNP ID
rs141703374
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67863680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGATCCCACCACTGCGAATGCT[C/T]GTCTGCTGGGGTGGAACCCACCTCC
Phenotype
MIM: 606030 MIM: 605813
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
EDC4 PubMed Links

Gene Details

Gene
EDC4
Gene Name
enhancer of mRNA decapping 4
There are no transcripts associated with this gene.

Gene
NUTF2
Gene Name
nuclear transport factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322038.1 Intron NP_001308967.1
NM_001322039.1 Intron NP_001308968.1
NM_001322040.1 Intron NP_001308969.1
NM_001322041.1 Intron NP_001308970.1
NM_005796.2 Intron NP_005787.1

View Full Product Details