Product Details

SNP ID
rs141749443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3658827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAGCTGAGCCAGGCCAGGCTCG[C/G]TTGCGCGCAGCTGATTCAGCTTCTT
Phenotype
MIM: 125505 MIM: 606219
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNASE1 PubMed Links

Gene Details

Gene
DNASE1
Gene Name
deoxyribonuclease I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005223.3 2068 Intron NP_005214.2
XM_005255148.3 2068 Intron XP_005255205.1
XM_006720854.3 2068 Intron XP_006720917.1
XM_011522393.2 2068 Intron XP_011520695.1
XM_017022992.1 2068 Intron XP_016878481.1
XM_017022993.1 2068 Intron XP_016878482.1
XM_017022994.1 2068 Intron XP_016878483.1
XM_017022995.1 2068 Intron XP_016878484.1
XM_017022996.1 2068 Intron XP_016878485.1
XM_017022997.1 2068 Intron XP_016878486.1
XM_017022998.1 2068 Intron XP_016878487.1
XM_017022999.1 2068 Intron XP_016878488.1
XM_017023000.1 2068 Intron XP_016878489.1
XM_017023001.1 2068 Intron XP_016878490.1
XM_017023002.1 2068 Intron XP_016878491.1
XM_017023003.1 2068 Intron XP_016878492.1
XM_017023004.1 2068 Intron XP_016878493.1
XM_017023005.1 2068 Intron XP_016878494.1
XM_017023006.1 2068 Intron XP_016878495.1
XM_017023007.1 2068 Intron XP_016878496.1
Gene
TRAP1
Gene Name
TNF receptor associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272049.1 2068 Missense Mutation ACC,AGC T607S NP_001258978.1
NM_016292.2 2068 Missense Mutation ACC,AGC T660S NP_057376.2
XM_011522345.1 2068 Missense Mutation ACC,AGC T520S XP_011520647.1
XM_017022851.1 2068 Missense Mutation ACC,AGC T361S XP_016878340.1

View Full Product Details