Product Details

SNP ID
rs142084682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67885790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGACACCATATACCAGGGCTTC[A/G]CCGAGTGTCTCATCCGCTTGGGGGA
Phenotype
MIM: 606030 MIM: 177015
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDC4 PubMed Links

Gene Details

Gene
EDC4
Gene Name
enhancer of mRNA decapping 4
There are no transcripts associated with this gene.

Gene
NRN1L
Gene Name
neuritin 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198443.1 174 Missense Mutation ACC,GCC T50A NP_940845.1
Gene
PSKH1
Gene Name
protein serine kinase H1
There are no transcripts associated with this gene.

View Full Product Details