Product Details
- SNP ID
-
rs142106374
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:1510979 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCTGCCTCTTCCACCACCTCCT[C/T]GTCCAGCTCCCTGGCGTCCTCCATG
- Phenotype
-
MIM: 614620
MIM: 611140
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IFT140
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1053730] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IFT140
- Gene Name
- intraflagellar transport 140
- Gene
- TELO2
- Gene Name
- telomere maintenance 2
View Full Product Details