Product Details

SNP ID
rs142138764
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81053634 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCCCCTCCACTAACTTTTCAT[A/T]CTTTACTTGGAAGCAAGTTACTATT
Phenotype
MIM: 614693
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATMIN PubMed Links

Gene Details

Gene
ATMIN
Gene Name
ATM interactor
There are no transcripts associated with this gene.

Gene
C16orf46
Gene Name
chromosome 16 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100873.1 2124 UTR 3 NP_001094343.1
NM_152337.2 2124 Intron NP_689550.2
XM_005255792.2 2124 UTR 3 XP_005255849.1
XM_011522848.2 2124 UTR 3 XP_011521150.2
XM_011522849.2 2124 UTR 3 XP_011521151.2
XM_017022916.1 2124 UTR 3 XP_016878405.1
XM_017022917.1 2124 UTR 3 XP_016878406.1

View Full Product Details