Product Details

SNP ID
rs142166852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2243215 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCCGATGGTGTTCTCCAGGGT[C/G]TCTTTCAACCTGGAAATGCAGACAC
Phenotype
MIM: 602622 MIM: 603022 MIM: 600305 MIM: 606447
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNASE1L2 PubMed Links

Gene Details

Gene
DNASE1L2
Gene Name
deoxyribonuclease I-like 2
There are no transcripts associated with this gene.

Gene
E4F1
Gene Name
E4F transcription factor 1
There are no transcripts associated with this gene.

Gene
ECI1
Gene Name
enoyl-CoA delta isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178029.1 608 Missense Mutation GAC,GAG D174E NP_001171500.1
NM_001919.3 608 Missense Mutation GAC,GAG D191E NP_001910.2
Gene
RNPS1
Gene Name
RNA binding protein with serine rich domain 1
There are no transcripts associated with this gene.

View Full Product Details