Product Details

SNP ID
rs142221190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30778725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAAGTTCCCATCTGACCTCCA[C/T]GTCCATAGCAGTTTCAAGTTTCACT
Phenotype
MIM: 607700
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF40 PubMed Links

Gene Details

Gene
RNF40
Gene Name
ring finger protein 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207033.1 5811 Intron NP_001193962.1
NM_001207034.1 5811 Intron NP_001193963.1
NM_001286572.2 5811 Intron NP_001273501.1
NM_014771.3 5811 Intron NP_055586.1
XM_011545997.1 5811 Intron XP_011544299.1
Gene
ZNF629
Gene Name
zinc finger protein 629
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080417.1 5811 UTR 3 NP_001073886.1
XM_005255222.3 5811 UTR 3 XP_005255279.1

View Full Product Details