Product Details

SNP ID
rs142275854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:651731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTGTGTCTCCAGTGGTCCAGA[C/T]GCCCAGCCCCACAGCCGTGAGTACC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371038 PubMed Links

Gene Details

Gene
LOC105371038
Gene Name
uncharacterized LOC105371038
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
There are no transcripts associated with this gene.

Gene
WDR90
Gene Name
WD repeat domain 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145294.4 878 Missense Mutation ACG,ATG T275M NP_660337.3
XM_017023023.1 878 Missense Mutation ACG,ATG T275M XP_016878512.1
XM_017023024.1 878 Intron XP_016878513.1

View Full Product Details