Product Details

SNP ID
rs142285591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29819705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACCCCAGCCCCGCCAGCCCCC[C/G]ACTCCGATCCTCCGGGAGTAGTCTC
Phenotype
MIM: 600999 MIM: 605088 MIM: 612033 MIM: 614386
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAZ PubMed Links

Gene Details

Gene
MAZ
Gene Name
MYC associated zinc finger protein
There are no transcripts associated with this gene.

Gene
MVP
Gene Name
major vault protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293204.1 1035 Intron NP_001280133.1
NM_001293205.1 1035 Intron NP_001280134.1
NM_005115.4 1035 Intron NP_005106.2
NM_017458.3 1035 Intron NP_059447.2
Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024516.3 1035 Missense Mutation CCA,CGA P239R NP_078792.1
Gene
PRRT2
Gene Name
proline rich transmembrane protein 2
There are no transcripts associated with this gene.

View Full Product Details