Product Details

SNP ID
rs142399311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31485758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCGTGGTGCTGCTACTGGGCTG[A/G]CTGTTTGCACCCGTGTACCTGACAG
Phenotype
MIM: 182381 MIM: 602353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf58 PubMed Links

Gene Details

Gene
C16orf58
Gene Name
chromosome 16 open reading frame 58
There are no transcripts associated with this gene.

Gene
LOC105371171
Gene Name
uncharacterized LOC105371171
There are no transcripts associated with this gene.

Gene
SLC5A2
Gene Name
solute carrier family 5 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003041.3 935 Nonsense Mutation TGA,TGG *111W NP_003032.1
XM_006721072.3 935 Nonsense Mutation TGA,TGG *118W XP_006721135.2
Gene
TGFB1I1
Gene Name
transforming growth factor beta 1 induced transcript 1
There are no transcripts associated with this gene.

View Full Product Details