Product Details

SNP ID
rs142477564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3582414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGCTTCTCCCTGCGGGTGGC[A/G]GCAGTGGTGAAGGTGATACAGTGGG
Phenotype
MIM: 615648 MIM: 613278
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NLRC3 PubMed Links

Gene Details

Gene
NLRC3
Gene Name
NLR family CARD domain containing 3
There are no transcripts associated with this gene.

Gene
SLX4
Gene Name
SLX4 structure-specific endonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032444.2 5465 Silent Mutation GCC,GCT A1811A NP_115820.2
XM_011522715.2 5465 Silent Mutation GCC,GCT A1810A XP_011521017.1
XM_017023775.1 5465 Silent Mutation GCC,GCT A1537A XP_016879264.1

View Full Product Details