Product Details
- SNP ID
-
rs142490370
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57759792 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTGGTCCCAGAGCTGGGGGCTGA[A/G]TGGGCCCGTGCCGAGGGCTGTGGCG
- Phenotype
-
MIM: 602703
MIM: 604535
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KATNB1
PubMed Links
Gene Details
- Gene
- KATNB1
- Gene Name
- katanin regulatory subunit B1
There are no transcripts associated with this gene.
- Gene
- KIFC3
- Gene Name
- kinesin family member C3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130099.1 |
3165 |
Silent Mutation |
CAC,CAT |
H665H |
NP_001123571.1 |
NM_001130100.1 |
3165 |
Silent Mutation |
CAC,CAT |
H804H |
NP_001123572.1 |
NM_001318710.1 |
3165 |
Silent Mutation |
CAC,CAT |
H826H |
NP_001305639.1 |
NM_001318711.1 |
3165 |
Silent Mutation |
CAC,CAT |
H746H |
NP_001305640.1 |
NM_001318712.1 |
3165 |
Silent Mutation |
CAC,CAT |
H702H |
NP_001305641.1 |
NM_001318713.1 |
3165 |
Silent Mutation |
CAC,CAT |
H662H |
NP_001305642.1 |
NM_001318714.1 |
3165 |
Silent Mutation |
CAC,CAT |
H665H |
NP_001305643.1 |
NM_001318715.1 |
3165 |
Silent Mutation |
CAC,CAT |
H665H |
NP_001305644.1 |
NM_005550.3 |
3165 |
Silent Mutation |
CAC,CAT |
H804H |
NP_005541.3 |
XM_005255937.1 |
3165 |
Silent Mutation |
CAC,CAT |
H967H |
XP_005255994.1 |
XM_005255944.1 |
3165 |
Silent Mutation |
CAC,CAT |
H746H |
XP_005256001.1 |
XM_006721188.1 |
3165 |
Silent Mutation |
CAC,CAT |
H993H |
XP_006721251.1 |
XM_011523075.1 |
3165 |
Silent Mutation |
CAC,CAT |
H853H |
XP_011521377.1 |
XM_011523076.1 |
3165 |
Silent Mutation |
CAC,CAT |
H853H |
XP_011521378.1 |
XM_011523077.1 |
3165 |
Silent Mutation |
CAC,CAT |
H832H |
XP_011521379.1 |
XM_011523078.1 |
3165 |
Silent Mutation |
CAC,CAT |
H665H |
XP_011521380.1 |
XM_011523079.1 |
3165 |
Silent Mutation |
CAC,CAT |
H665H |
XP_011521381.1 |
XM_017023219.1 |
3165 |
Silent Mutation |
CAC,CAT |
H1050H |
XP_016878708.1 |
XM_017023220.1 |
3165 |
Silent Mutation |
CAC,CAT |
H1028H |
XP_016878709.1 |
XM_017023221.1 |
3165 |
Silent Mutation |
CAC,CAT |
H967H |
XP_016878710.1 |
XM_017023222.1 |
3165 |
Silent Mutation |
CAC,CAT |
H883H |
XP_016878711.1 |
XM_017023223.1 |
3165 |
Silent Mutation |
CAC,CAT |
H826H |
XP_016878712.1 |
XM_017023224.1 |
3165 |
Silent Mutation |
CAC,CAT |
H883H |
XP_016878713.1 |
XM_017023225.1 |
3165 |
Silent Mutation |
CAC,CAT |
H747H |
XP_016878714.1 |
XM_017023226.1 |
3165 |
Silent Mutation |
CAC,CAT |
H746H |
XP_016878715.1 |
XM_017023227.1 |
3165 |
Silent Mutation |
CAC,CAT |
H746H |
XP_016878716.1 |
View Full Product Details