Product Details

SNP ID
rs142697731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46692509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTAGCTGTACAGTTTAGCTGTA[C/T]AGAAGCAGTGAACATGGCTTCAAAG
Phenotype
MIM: 612147 MIM: 607213 MIM: 601501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYLK3 PubMed Links

Gene Details

Gene
MYLK3
Gene Name
myosin light chain kinase 3
There are no transcripts associated with this gene.

Gene
ORC6
Gene Name
origin recognition complex subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014321.3 373 Missense Mutation ACA,ATA T108I NP_055136.1
XM_011522978.2 373 Missense Mutation ACA,ATA T108I XP_011521280.1
Gene
VPS35
Gene Name
VPS35 retromer complex component
There are no transcripts associated with this gene.

View Full Product Details