Product Details

SNP ID
rs142710856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30359306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCGCAGGTGGCGATGCGCCAGC[A/G]GGGAGGCCCGGCGCAGGAGTGCAAA
Phenotype
MIM: 604470 MIM: 613620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD2BP2 PubMed Links

Gene Details

Gene
CD2BP2
Gene Name
CD2 cytoplasmic tail binding protein 2
There are no transcripts associated with this gene.

Gene
TBC1D10B
Gene Name
TBC1 domain family member 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015527.3 1526 Missense Mutation CCG,CTG P503L NP_056342.3
XM_011545789.1 1526 Missense Mutation CCG,CTG P398L XP_011544091.1
XM_011545790.2 1526 Intron XP_011544092.1

View Full Product Details