Product Details

SNP ID
rs142869246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1979522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCGCGGCTGCTCTCGTAGGCGC[A/G]GGAAGCACAGAACTGGGGACCTGGT
Phenotype
MIM: 600924 MIM: 611256 MIM: 605915
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 1103 Missense Mutation CGC,TGC R245C NP_001254650.1
NM_144603.3 1103 Missense Mutation CGC,TGC R240C NP_653204.1
NM_172167.2 1103 Missense Mutation CGC,TGC R241C NP_751907.1
NM_172168.2 1103 Missense Mutation CGC,TGC R246C NP_751908.1
XM_005255099.4 1103 Missense Mutation CGC,TGC R246C XP_005255156.1
XM_017022927.1 1103 Missense Mutation CGC,TGC R246C XP_016878416.1
XM_017022928.1 1103 Missense Mutation CGC,TGC R246C XP_016878417.1
Gene
TBL3
Gene Name
transducin beta like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006453.2 1103 Intron NP_006444.2

View Full Product Details