Product Details

SNP ID
rs143203977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57252144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAGGGCTCTTTCTAGTCCTTC[C/G]TTTGGTTGTTTTCTCATATAGGAAA
Phenotype
MIM: 615407 MIM: 600340
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARL2BP PubMed Links

Gene Details

Gene
ARL2BP
Gene Name
ADP ribosylation factor like GTPase 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012106.3 Intron NP_036238.1
XM_011522977.1 Intron XP_011521279.1
Gene
PLLP
Gene Name
plasmolipin
There are no transcripts associated with this gene.

View Full Product Details