Product Details

SNP ID
rs143274002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71536797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCTGTCTATGAAGGCACAGCC[C/T]GAGCGCATGCACGTGCTGGTTCTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST4 PubMed Links

Gene Details

Gene
CHST4
Gene Name
carbohydrate sulfotransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166395.1 331 Silent Mutation CCC,CCT P40P NP_001159867.1
NM_005769.2 331 Silent Mutation CCC,CCT P40P NP_005760.1

View Full Product Details