Product Details
- SNP ID
-
rs143292206
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81322021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACACTCTACCAGAGTTCCTGATTC[C/T]GGTGCTTGGAAGAGCAGAGTTAAAT
- Phenotype
-
MIM: 605379
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAN
PubMed Links
Gene Details
- Gene
- GAN
- Gene Name
- gigaxonin
View Full Product Details