Product Details

SNP ID
rs143437222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:74622897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAATGTATAAAGAACACTTAA[C/G]AACAGCCCCTCAGTCTGAGGAATTG
Phenotype
MIM: 614151
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RFWD3 PubMed Links

Gene Details

Gene
RFWD3
Gene Name
ring finger and WD repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018124.3 3407 UTR 3 NP_060594.3
XM_005256021.4 3407 UTR 3 XP_005256078.1
XM_005256022.4 3407 UTR 3 XP_005256079.1
XM_006721228.3 3407 UTR 3 XP_006721291.1
XM_011523191.2 3407 UTR 3 XP_011521493.1
XM_017023391.1 3407 UTR 3 XP_016878880.1
XM_017023392.1 3407 UTR 3 XP_016878881.1

View Full Product Details