Product Details

SNP ID
rs143552960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:10528918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCATCATCAGTGAACACAGAGC[A/G]TCCTAGAGCGCCAGCCTGTTCATGA
Phenotype
MIM: 602334
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMP2 PubMed Links

Gene Details

Gene
EMP2
Gene Name
epithelial membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001424.5 4713 UTR 3 NP_001415.1
XM_006720864.3 4713 UTR 3 XP_006720927.1

View Full Product Details