Product Details

SNP ID
rs143704082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67483270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCCCACCCTTGCTCACACTGACC[C/T]GGGAGCTCTGGGAGCAGGGCCTGGT
Phenotype
MIM: 602311 MIM: 607028
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGRP PubMed Links

Gene Details

Gene
AGRP
Gene Name
agouti related neuropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001138.1 429 Silent Mutation CCA,CCG P43P NP_001129.1
Gene
ATP6V0D1
Gene Name
ATPase H+ transporting V0 subunit d1
There are no transcripts associated with this gene.

View Full Product Details