Product Details

SNP ID
rs144067732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67390339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGCTGGTGTCCGTCAGCCGGTC[C/T]ACAGCACCCCCTGTTTTTGCTTTCT
Phenotype
MIM: 616957
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC36 PubMed Links

Gene Details

Gene
LRRC36
Gene Name
leucine rich repeat containing 36
There are no transcripts associated with this gene.

Gene
TPPP3
Gene Name
tubulin polymerization promoting protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015964.3 528 Silent Mutation GTA,GTG V122V NP_057048.2
NM_016140.3 528 Silent Mutation GTA,GTG V122V NP_057224.2
Gene
ZDHHC1
Gene Name
zinc finger DHHC-type containing 1
There are no transcripts associated with this gene.

View Full Product Details