Product Details

SNP ID
rs144127842
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:681811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCAGGGAGCTGGAAGAGTGCCA[G/T]CGAAACCACGAGGGTGATGAGGACG
Phenotype
MIM: 603264 MIM: 613889 MIM: 607207
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
JMJD8 PubMed Links
Additional Information
For this assay, SNP(s) [rs1046112] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
JMJD8
Gene Name
jumonji domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005920.2 1900 UTR 3 NP_001005920.2
NM_001323918.1 1900 UTR 3 NP_001310847.1
NM_001323919.1 1900 UTR 3 NP_001310848.1
NM_001323920.1 1900 UTR 3 NP_001310849.1
NM_001323922.1 1900 UTR 3 NP_001310851.1
Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
There are no transcripts associated with this gene.

Gene
RHOT2
Gene Name
ras homolog family member T2
There are no transcripts associated with this gene.

Gene
STUB1
Gene Name
STIP1 homology and U-box containing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293197.1 1900 Silent Mutation CAG,CAT Q109H NP_001280126.1
NM_005861.3 1900 Silent Mutation CAG,CAT Q181H NP_005852.2
Gene
WDR24
Gene Name
WD repeat domain 24
There are no transcripts associated with this gene.

View Full Product Details