Product Details

SNP ID
rs144130252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31473899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCCCTCCATTCTCCTCTTCC[A/G]GCGGTGTCTTGGGTACCGGGCTCTG
Phenotype
MIM: 615549 MIM: 182381 MIM: 602353
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMC5 PubMed Links

Gene Details

Gene
ARMC5
Gene Name
armadillo repeat containing 5
There are no transcripts associated with this gene.

Gene
SLC5A2
Gene Name
solute carrier family 5 member 2
There are no transcripts associated with this gene.

Gene
TGFB1I1
Gene Name
transforming growth factor beta 1 induced transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042454.2 250 Missense Mutation AGC,GGC S83G NP_001035919.1
NM_001164719.1 250 Missense Mutation AGC,GGC S66G NP_001158191.1
NM_015927.4 250 Missense Mutation AGC,GGC S66G NP_057011.2

View Full Product Details