Product Details
- SNP ID
-
rs144133911
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71648914 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCATAGTGCTGTGTCGAACTCCTC[C/T]GGGGGCTCGGTCCGATCCTCTTCAT
- Phenotype
-
MIM: 614094
MIM: 611066
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MARVELD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61733125] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARVELD3
- Gene Name
- MARVEL domain containing 3
There are no transcripts associated with this gene.
- Gene
- PHLPP2
- Gene Name
- PH domain and leucine rich repeat protein phosphatase 2
View Full Product Details