Product Details

SNP ID
rs144276368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22026176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACAATTATTTATGGTCTGTATG[A/G]TAACTAAGTCTGGAAGTTGGATATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf52 PubMed Links

Gene Details

Gene
C16orf52
Gene Name
chromosome 16 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164579.1 Intron NP_001158051.1

View Full Product Details