Product Details

SNP ID
rs144503112
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:21199626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAACACAGTGGGGAGTCAGGG[G/T]AGAGTCGATTACAGATTAAGGCACT
Phenotype
MIM: 182888
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ZP2 PubMed Links

Gene Details

Gene
ZP2
Gene Name
zona pellucida glycoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290104.1 2327 Missense Mutation NP_001277033.1
NM_003460.2 2327 Missense Mutation NP_003451.1
XM_011545942.2 2327 Missense Mutation XP_011544244.1

View Full Product Details