Product Details

SNP ID
rs144633852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66910038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGTGGCTGACCACCACGGGGAT[C/T]ATGTGTTCACGTGGCTCCACCCAAT
Phenotype
MIM: 603118
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH16 PubMed Links

Gene Details

Gene
CDH16
Gene Name
cadherin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204744.1 2308 Missense Mutation ATA,ATG I719M NP_001191673.1
NM_001204745.1 2308 Missense Mutation ATA,ATG I702M NP_001191674.1
NM_001204746.1 2308 Missense Mutation ATA,ATG I644M NP_001191675.1
NM_004062.3 2308 Missense Mutation ATA,ATG I741M NP_004053.1
XM_005255770.2 2308 Missense Mutation ATA,ATG I563M XP_005255827.1
XM_011522807.1 2308 Missense Mutation ATA,ATG I678M XP_011521109.1

View Full Product Details