Product Details

SNP ID
rs144739067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30524667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGAGGAGCGGCAGAAGCCCTT[A/G]CCGCACACGGCGCACCTGTAGGGCC
Phenotype
MIM: 153370
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGAL PubMed Links

Gene Details

Gene
ITGAL
Gene Name
integrin subunit alpha L
There are no transcripts associated with this gene.

Gene
ZNF747
Gene Name
zinc finger protein 747
There are no transcripts associated with this gene.

Gene
ZNF768
Gene Name
zinc finger protein 768
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024671.3 1474 Silent Mutation GGC,GGT G491G NP_078947.3
XM_017023665.1 1474 Silent Mutation GGC,GGT G515G XP_016879154.1
XM_017023666.1 1474 Silent Mutation GGC,GGT G460G XP_016879155.1

View Full Product Details