Product Details

SNP ID
rs144830689
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:47082304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACCAACAGAATGGGAGCTATG[C/T]AACAATCTAGCGAAGAGAAACATTG
Phenotype
MIM: 607974
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NETO2 PubMed Links

Gene Details

Gene
NETO2
Gene Name
neuropilin and tolloid like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201477.1 2887 UTR 3 NP_001188406.1
NM_018092.4 2887 UTR 3 NP_060562.3
XM_006721289.1 2887 UTR 3 XP_006721352.1
XM_006721291.1 2887 UTR 3 XP_006721354.1
XM_006721292.1 2887 Intron XP_006721355.1
XM_017023740.1 2887 UTR 3 XP_016879229.1

View Full Product Details