Product Details

SNP ID
rs145062714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72787459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAGGCGTGGGGGGAAGCGGAG[G/T]AGGGGGCGGCGGCCGACGGGGGAGG
Phenotype
MIM: 104155
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFHX3 PubMed Links

Gene Details

Gene
ZFHX3
Gene Name
zinc finger homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164766.1 8204 Missense Mutation TAC,TCC Y2692S NP_001158238.1
NM_006885.3 8204 Missense Mutation TAC,TCC Y3606S NP_008816.3
XM_005255957.3 8204 Missense Mutation TAC,TCC Y3605S XP_005256014.1
XM_017023251.1 8204 Missense Mutation TAC,TCC Y2723S XP_016878740.1

View Full Product Details