Product Details

SNP ID
rs145081403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4461789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCGAGGCGGGCAGGTCACAGCAG[A/G]TTGAAGTCCCCTTTGTGCTGTTCCA
Phenotype
MIM: 608382
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJA3 PubMed Links

Gene Details

Gene
DNAJA3
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
There are no transcripts associated with this gene.

Gene
NMRAL1
Gene Name
NmrA-like family domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305141.1 1204 Silent Mutation NP_001292070.1
NM_001305142.1 1204 Silent Mutation NP_001292071.1
NM_020677.4 1204 Silent Mutation NP_065728.1
XM_006720905.3 1204 Silent Mutation XP_006720968.1
XM_006720906.2 1204 Silent Mutation XP_006720969.1
XM_011522567.2 1204 Intron XP_011520869.1
XM_017023485.1 1204 Silent Mutation XP_016878974.1
XM_017023486.1 1204 Intron XP_016878975.1
XM_017023487.1 1204 Intron XP_016878976.1
XM_017023488.1 1204 Intron XP_016878977.1
XM_017023489.1 1204 Silent Mutation XP_016878978.1
XM_017023490.1 1204 Silent Mutation XP_016878979.1
XM_017023491.1 1204 UTR 3 XP_016878980.1
XM_017023492.1 1204 UTR 3 XP_016878981.1

View Full Product Details