Product Details

SNP ID
rs145313679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1361902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATGAGTTCTGCCCGTTCCACAA[C/T]GTGACCCAGCACGAGCAGACCTTCC
Phenotype
MIM: 607838
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GNPTG PubMed Links

Gene Details

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032520.4 347 Silent Mutation AAC,AAT N88N NP_115909.1
XM_017023782.1 347 Silent Mutation AAC,AAT N104N XP_016879271.1
XM_017023783.1 347 UTR 5 XP_016879272.1
Gene
UNKL
Gene Name
unkempt family like zinc finger
There are no transcripts associated with this gene.

View Full Product Details