Product Details

SNP ID
rs145386234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30525580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGGGTGAACTCCTACGGAGATA[C/T]TCAAAGGACTCTTTTCCTCGGGGTT
Phenotype
MIM: 153370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ITGAL PubMed Links

Gene Details

Gene
ITGAL
Gene Name
integrin subunit alpha L
There are no transcripts associated with this gene.

Gene
ZNF747
Gene Name
zinc finger protein 747
There are no transcripts associated with this gene.

Gene
ZNF768
Gene Name
zinc finger protein 768
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024671.3 561 Missense Mutation AAT,AGT N187S NP_078947.3
XM_017023665.1 561 Missense Mutation AAT,AGT N211S XP_016879154.1
XM_017023666.1 561 Missense Mutation AAT,AGT N156S XP_016879155.1

View Full Product Details