Product Details

SNP ID
rs145422527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15434752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGGCTTGGAAAACGGAGAGACT[A/G]GGGAGAAGTGAGTACTGGGGCGGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf45 PubMed Links

Gene Details

Gene
C16orf45
Gene Name
chromosome 16 open reading frame 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142469.1 285 Intron NP_001135941.1
NM_033201.2 285 Silent Mutation CTA,CTG L33L NP_149978.1
XM_011522731.2 285 Silent Mutation CTA,CTG L33L XP_011521033.1
XM_017023824.1 285 UTR 5 XP_016879313.1

View Full Product Details