Product Details

SNP ID
rs145458736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15596543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCTCAAAGCTGTGTTCAATGG[A/C]AAAGAAAAACATGATAGAACACAGG
Phenotype
MIM: 614593
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C16orf45 PubMed Links

Gene Details

Gene
C16orf45
Gene Name
chromosome 16 open reading frame 45
There are no transcripts associated with this gene.

Gene
KIAA0430
Gene Name
KIAA0430
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184998.1 5420 UTR 3 NP_001171927.1
NM_001184999.1 5420 UTR 3 NP_001171928.1
NM_014647.3 5420 UTR 3 NP_055462.2
XM_005255704.1 5420 UTR 3 XP_005255761.1
XM_005255706.1 5420 UTR 3 XP_005255763.1
XM_005255707.1 5420 UTR 3 XP_005255764.1
XM_017023900.1 5420 UTR 3 XP_016879389.1
XM_017023901.1 5420 UTR 3 XP_016879390.1
XM_017023902.1 5420 UTR 3 XP_016879391.1
XM_017023903.1 5420 UTR 3 XP_016879392.1
XM_017023904.1 5420 UTR 3 XP_016879393.1
XM_017023905.1 5420 UTR 3 XP_016879394.1
XM_017023906.1 5420 UTR 3 XP_016879395.1

View Full Product Details