Product Details

SNP ID
rs145505967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30353791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGCTGGCCAGGTGGCCGCTCCC[A/G]GATCTTCACCTGTAATGGGAAGATG
Phenotype
MIM: 604470 MIM: 613620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD2BP2 PubMed Links

Gene Details

Gene
CD2BP2
Gene Name
CD2 cytoplasmic tail binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243646.1 561 Silent Mutation NP_001230575.1
NM_006110.2 561 Silent Mutation NP_006101.1
Gene
TBC1D10B
Gene Name
TBC1 domain family member 10B
There are no transcripts associated with this gene.

View Full Product Details