Product Details

SNP ID
rs145571848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30358055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAAAGCTTCCGGCCTTGAGCCTT[A/C]TTCTCCTGCTTCTGCCGCTCCTTCT
Phenotype
MIM: 604470 MIM: 613620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD2BP2 PubMed Links

Gene Details

Gene
CD2BP2
Gene Name
CD2 cytoplasmic tail binding protein 2
There are no transcripts associated with this gene.

Gene
TBC1D10B
Gene Name
TBC1 domain family member 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015527.3 2201 Missense Mutation AAG,AAT K772N NP_056342.3
XM_011545789.1 2201 Missense Mutation AAG,AAT K667N XP_011544091.1
XM_011545790.2 2201 UTR 3 XP_011544092.1

View Full Product Details