Product Details

SNP ID
rs145637964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:590329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTCCGGTGAAGAGCTACGACT[A/T]CCTGCTCAAGTTCCTGCTGGTGGGC
Phenotype
MIM: 605754
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929280 PubMed Links

Gene Details

Gene
LOC101929280
Gene Name
uncharacterized LOC101929280
There are no transcripts associated with this gene.

Gene
PIGQ
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Q
There are no transcripts associated with this gene.

Gene
RAB40C
Gene Name
RAB40C, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172663.1 112 Missense Mutation TAC,TTC Y13F NP_001166134.1
NM_001172664.1 112 Missense Mutation TAC,TTC Y13F NP_001166135.1
NM_001172665.1 112 Missense Mutation TAC,TTC Y13F NP_001166136.1
NM_001172666.1 112 Missense Mutation TAC,TTC Y13F NP_001166137.1
NM_021168.4 112 Missense Mutation TAC,TTC Y13F NP_066991.3

View Full Product Details