Product Details

SNP ID
rs145932509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66723775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACCATAGAGTCTGGCTTTCGAG[C/T]CATTCTATCCAGTTCTTCCTGAACA
Phenotype
MIM: 611406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DYNC1LI2 PubMed Links

Gene Details

Gene
DYNC1LI2
Gene Name
dynein cytoplasmic 1 light intermediate chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286157.1 1457 Missense Mutation ACT,GCT T399A NP_001273086.1
NM_001323955.1 1457 Missense Mutation ACT,GCT T437A NP_001310884.1
NM_006141.2 1457 Missense Mutation ACT,GCT T476A NP_006132.1
XM_017023008.1 1457 Intron XP_016878497.1

View Full Product Details