Product Details

SNP ID
rs146083652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57465720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATGATATTCTATAAGTCAGTGT[G/T]TTCTGTGAAGCAGGGAGCCCCCTGC
Phenotype
MIM: 612837 MIM: 608333 MIM: 180663
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COQ9 PubMed Links

Gene Details

Gene
COQ9
Gene Name
coenzyme Q9
There are no transcripts associated with this gene.

Gene
DOK4
Gene Name
docking protein 4
There are no transcripts associated with this gene.

Gene
POLR2C
Gene Name
RNA polymerase II subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032940.2 Intron NP_116558.1

View Full Product Details