Product Details

SNP ID
rs146411391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72120043 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTTCTCTTCTATCTCCTTCTTCA[A/G]CTTCTTGTTTTCCTGCTGCAAGTGG
Phenotype
MIM: 605584
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX38 PubMed Links

Gene Details

Gene
DHX38
Gene Name
DEAH-box helicase 38
There are no transcripts associated with this gene.

Gene
PMFBP1
Gene Name
polyamine modulated factor 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160213.1 2897 Silent Mutation CTG,TTG L814L NP_001153685.1
NM_031293.2 2897 Silent Mutation CTG,TTG L939L NP_112583.2
XM_011523357.2 2897 Silent Mutation CTG,TTG L964L XP_011521659.1
XM_011523358.2 2897 Silent Mutation CTG,TTG L964L XP_011521660.1
XM_011523360.2 2897 Silent Mutation CTG,TTG L964L XP_011521662.1
XM_011523361.2 2897 Silent Mutation CTG,TTG L964L XP_011521663.1
XM_011523362.2 2897 Silent Mutation CTG,TTG L961L XP_011521664.1

View Full Product Details