Product Details

SNP ID
rs146485007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25217022 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCAGCAGCTCAGGCAAGAGTC[C/T]GATGTTTGTGCCATCTGATCCTGAT
Phenotype
MIM: 603750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP8 PubMed Links

Gene Details

Gene
AQP8
Gene Name
aquaporin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169.2 59 UTR 5 NP_001160.2
XM_011545822.2 59 UTR 5 XP_011544124.1
XM_011545823.2 59 UTR 5 XP_011544125.1

View Full Product Details