Product Details

SNP ID
rs146497747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23064023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAGTTAAAGACAGCCTGCACAG[C/T]AACAGCTACTTTTTGTGTTCGTTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP31 PubMed Links

Gene Details

Gene
USP31
Gene Name
ubiquitin specific peptidase 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020718.3 7199 UTR 3 NP_065769.3
XM_017023496.1 7199 UTR 3 XP_016878985.1
XM_017023497.1 7199 UTR 3 XP_016878986.1
XM_017023498.1 7199 UTR 3 XP_016878987.1
XM_017023499.1 7199 UTR 3 XP_016878988.1
XM_017023500.1 7199 Intron XP_016878989.1

View Full Product Details