Product Details
- SNP ID
-
rs146555912
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:31722695 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGTGGGAACACCTGGACTCAGAT[C/G]AGAAGCTTTTATATGGGGATGTGAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF720
PubMed Links
Gene Details
- Gene
- ZNF720
- Gene Name
- zinc finger protein 720
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130913.1 |
277 |
Missense Mutation |
CAG,GAG |
Q25E |
NP_001124385.1 |
View Full Product Details