Product Details
- SNP ID
-
rs146601305
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:28824878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGTGTTTGCTGGCTTATTAATGT[C/T]TATTTTTTTCTTCCTCTACAAGGTG
- Phenotype
-
MIM: 607931
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN2L
PubMed Links
Gene Details
- Gene
- ATXN2L
- Gene Name
- ataxin 2 like
View Full Product Details