Product Details

SNP ID
rs146953404
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:53704652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTGGAACAATTGTCTCCGTG[A/G]CCGGCTGATAACAGTTTGTGTCTTT
Phenotype
MIM: 610966 MIM: 610937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTO PubMed Links

Gene Details

Gene
FTO
Gene Name
fat mass and obesity associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080432.2 Intron NP_001073901.1
XM_011523313.2 Intron XP_011521615.1
XM_011523314.2 Intron XP_011521616.1
XM_011523315.2 Intron XP_011521617.1
XM_011523316.2 Intron XP_011521618.1
XM_017023654.1 Intron XP_016879143.1
XM_017023655.1 Intron XP_016879144.1
XM_017023656.1 Intron XP_016879145.1
XM_017023657.1 Intron XP_016879146.1
XM_017023658.1 Intron XP_016879147.1
Gene
RPGRIP1L
Gene Name
RPGRIP1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005255867.1 Intron XP_005255924.1
XM_005255868.1 Intron XP_005255925.1
XM_011522970.1 Intron XP_011521272.1
XM_011522971.2 Intron XP_011521273.1
XM_011522973.2 Intron XP_011521275.1
XM_017023094.1 Intron XP_016878583.1
XM_017023095.1 Intron XP_016878584.1
XM_017023096.1 Intron XP_016878585.1
XM_017023097.1 Intron XP_016878586.1
XM_017023098.1 Intron XP_016878587.1
XM_017023099.1 Intron XP_016878588.1
XM_017023100.1 Intron XP_016878589.1

View Full Product Details