Product Details

SNP ID
rs147033584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2214031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCGCTGGGCGGCCATCTCCAC[A/G]GCTCGGACCAGACACCCGGTACCTG
Phenotype
MIM: 603022 MIM: 612190 MIM: 172280
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRICD5 PubMed Links

Gene Details

Gene
BRICD5
Gene Name
BRICHOS domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182563.3 707 Intron NP_872369.2
XM_005255263.4 707 Intron XP_005255320.1
Gene
E4F1
Gene Name
E4F transcription factor 1
There are no transcripts associated with this gene.

Gene
MLST8
Gene Name
MTOR associated protein, LST8 homolog
There are no transcripts associated with this gene.

Gene
PGP
Gene Name
phosphoglycolate phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042371.2 707 Silent Mutation GCC,GCT A221A NP_001035830.1

View Full Product Details