Product Details

SNP ID
rs147051608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57407197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCAGCAGAGGGAAAGGGGGTCA[C/T]GGGATTGGAGACAGTAAAGAGCTAG
Phenotype
MIM: 601520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL17 PubMed Links

Gene Details

Gene
CCL17
Gene Name
C-C motif chemokine ligand 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002987.2 Intron NP_002978.1
XM_011523256.2 Intron XP_011521558.1
XM_017023530.1 Intron XP_016879019.1

View Full Product Details