Product Details

SNP ID
rs147152324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66510057 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAATAAAAACACGTTAATCTAC[C/T]GGCAAAAGCAGCTGGCAGAAGCTAA
Phenotype
MIM: 188250
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TK2 PubMed Links

Gene Details

Gene
TK2
Gene Name
thymidine kinase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172643.1 2834 UTR 3 NP_001166114.1
NM_001172644.1 2834 UTR 3 NP_001166115.1
NM_001172645.1 2834 UTR 3 NP_001166116.1
NM_001271934.1 2834 UTR 3 NP_001258863.1
NM_001271935.1 2834 UTR 3 NP_001258864.1
NM_001272050.1 2834 UTR 3 NP_001258979.1
NM_004614.4 2834 UTR 3 NP_004605.4

View Full Product Details